ADVH49B82275 AChemBlock

2-Methoxymethylpyridine-3-boronic acid pinacol ester

  • Cas Number : 1430401-34-5
  • Molecular Weight : 267.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/2-methoxymethylpyridine-3-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70795
Availability details for
Quantity
  • Availability
  • Add to Cart