ADVH49B82279 AChemBlock

6-Chloro-5-nitropyridine-3-boronic acid pinacol ester

  • Cas Number : 1310383-11-9
  • Molecular Weight : 302.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-5-nitropyridine-3-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70635
Availability details for
Quantity
  • Availability
  • Add to Cart