ADVH49B8227E AChemBlock

trans-N,N,2-trimethylpyrrolidin-3-amine dihydrochloride

  • Molecular Weight : 201.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-n-n-2-trimethylpyrrolidin-3-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84349
Availability details for
Quantity
  • Availability
  • Add to Cart