ADVH49B82280 AChemBlock

trans-5-(trifluoromethyl)piperidin-3-ol

  • Molecular Weight : 169.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-5-trifluoromethyl-piperidin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84352
Availability details for
Quantity
  • Availability
  • Add to Cart