ADVH49B82281 AChemBlock

trans-3-bromocyclobutan-1-amine hydrochloride

  • Molecular Weight : 186.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-3-bromocyclobutan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84356
Availability details for
Quantity
  • Availability
  • Add to Cart