ADVH49B82286 AChemBlock

tert-butyl trans-4-(hydroxymethyl)-3-methoxy-piperidine-1-carboxylate

  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-trans-4-hydroxymethyl-3-methoxy-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84376
Availability details for
Quantity
  • Availability
  • Add to Cart