ADVH49B8228B AChemBlock

tert-butyl N-tert-butoxycarbonyl-N-[3-(hydroxymethyl)oxetan-3-yl]carbamate

  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-tert-butoxycarbonyl-n-3-hydroxymethyl-oxetan-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84386
Availability details for
Quantity
  • Availability
  • Add to Cart