ADVH49B8228E AChemBlock

tert-butyl N-[4-(aminomethyl)-2-methyl-cyclohexyl]carbamate

  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-aminomethyl-2-methyl-cyclohexyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84435
Availability details for
Quantity
  • Availability
  • Add to Cart