ADVH49B82290 AChemBlock

tert-butyl N-[3-(aminomethyl)oxetan-3-yl]carbamate hydrochloride

  • Molecular Weight : 238.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3-aminomethyl-oxetan-3-yl-carbamate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84439
Availability details for
Quantity
  • Availability
  • Add to Cart