ADVH49B82297 AChemBlock

tert-butyl N-[(8R)-6-oxa-2-azaspiro[3.4]octan-8-yl]carbamate

  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-8r-6-oxa-2-azaspiro-3-4-octan-8-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84451
Availability details for
Quantity
  • Availability
  • Add to Cart