ADVH49B82299 AChemBlock

tert-butyl N-[(3S,5S)-5-(difluoromethyl)-3-piperidyl]carbamate

  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3s-5s-5-difluoromethyl-3-piperidyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84459
Availability details for
Quantity
  • Availability
  • Add to Cart