ADVH49B8229E AChemBlock

tert-butyl N-[(1S,4R,6R)-2-azabicyclo[2.2.1]heptan-6-yl]carbamate

  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1s-4r-6r-2-azabicyclo-2-2-1-heptan-6-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84490
Availability details for
Quantity
  • Availability
  • Add to Cart