ADVH49B8229F AChemBlock

tert-butyl N-[(1R,2S,4S)-rel-4-amino-2-fluoro-cyclohexyl]carbamate

  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1r-2s-4s-rel-4-amino-2-fluoro-cyclohexyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84499
Availability details for
Quantity
  • Availability
  • Add to Cart