ADVH49B822AB AChemBlock

tert-butyl N-(8-methyl-3-azabicyclo[3.2.1]octan-8-yl)carbamate

  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-8-methyl-3-azabicyclo-3-2-1-octan-8-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84517
Availability details for
Quantity
  • Availability
  • Add to Cart