ADVH49B822B1 AChemBlock

tert-butyl N-(6-aminospiro[3.3]heptan-2-yl)carbamate hydrochloride

  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-6-aminospiro-3-3-heptan-2-yl-carbamate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84525
Availability details for
Quantity
  • Availability
  • Add to Cart