ADVH49B822B2 AChemBlock

tert-butyl N-(5-oxo-7-oxaspiro[3.5]nonan-2-yl)carbamate

  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-5-oxo-7-oxaspiro-3-5-nonan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84526
Availability details for
Quantity
  • Availability
  • Add to Cart