ADVH49B822B5 AChemBlock

tert-butyl N-(5-cyanonorpinan-1-yl)carbamate

  • Molecular Weight : 236.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-5-cyanonorpinan-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84530
Availability details for
Quantity
  • Availability
  • Add to Cart