ADVH49B822BB AChemBlock

tert-butyl N-(3-amino-1-oxaspiro[3.3]heptan-6-yl)carbamate

  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3-amino-1-oxaspiro-3-3-heptan-6-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84539
Availability details for
Quantity
  • Availability
  • Add to Cart