ADVH49B822BC AChemBlock

tert-butyl N-(2-azaspiro[4.5]decan-4-ylmethyl)carbamate

  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-azaspiro-4-5-decan-4-ylmethyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84540
Availability details for
Quantity
  • Availability
  • Add to Cart