ADVH49B822BD AChemBlock

tert-butyl N-(2-aminospiro[3.5]nonan-9-yl)carbamate

  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-aminospiro-3-5-nonan-9-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84541
Availability details for
Quantity
  • Availability
  • Add to Cart