ADVH49B822C3 AChemBlock

tert-butyl N-(2,3-dihydro-1H-pyrrolo[3,4-c]pyridin-6-yl)carbamate

  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-3-dihydro-1h-pyrrolo-3-4-c-pyridin-6-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84547
Availability details for
Quantity
  • Availability
  • Add to Cart