ADVH49B822C4 AChemBlock

tert-butyl N-(1,2,3,3a,4,5,6,6a-octahydrocyclopenta[b]pyrrol-5-yl)carbamate

  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1-2-3-3a-4-5-6-6a-octahydrocyclopenta-b-pyrrol-5-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84548
Availability details for
Quantity
  • Availability
  • Add to Cart