ADVH49B822C7 AChemBlock

tert-butyl 8-amino-4,4-difluoro-1,3-dihydroisoquinoline-2-carboxylate

  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-8-amino-4-4-difluoro-1-3-dihydroisoquinoline-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84573
Availability details for
Quantity
  • Availability
  • Add to Cart