ADVH49B822C8 AChemBlock

tert-butyl 7-oxo-2,3,4a,5,6,7a-hexahydro-1H-pyrrolo[3,4-b]pyrazine-4-carboxylate

  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-oxo-2-3-4a-5-6-7a-hexahydro-1h-pyrrolo-3-4-b-pyrazine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84576
Availability details for
Quantity
  • Availability
  • Add to Cart