ADVH49B822CC AChemBlock

tert-butyl 7-fluoro-5-(hydroxymethyl)-2-azaspiro[3.4]octane-2-carboxylate

  • Molecular Weight : 259.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-fluoro-5-hydroxymethyl-2-azaspiro-3-4-octane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84582
Availability details for
Quantity
  • Availability
  • Add to Cart