ADVH49B822D2 AChemBlock

tert-butyl 7,7-difluoro-3-iodo-6,8-dihydro-4H-pyrazolo[1,5-a][1,4]diazepine-5-carboxylate

  • Molecular Weight : 399.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-7-difluoro-3-iodo-6-8-dihydro-4h-pyrazolo-1-5-a-1-4-diazepine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84590
Availability details for
Quantity
  • Availability
  • Add to Cart