ADVH49B822D6 AChemBlock

tert-butyl 6-ethynyl-1,3-dihydropyrrolo[3,4-c]pyridine-2-carboxylate

  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-ethynyl-1-3-dihydropyrrolo-3-4-c-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84594
Availability details for
Quantity
  • Availability
  • Add to Cart