ADVH49B822DA AChemBlock

(2R,3S)-3-methylhex-5-ene-2-sulfonamide

  • Cas Number : 1638587-36-6
  • Molecular Weight : 177.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-3s-3-methylhex-5-ene-2-sulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78979
Availability details for
Quantity
  • Availability
  • Add to Cart