ADVH49B822DB AChemBlock

3-amino-4-nitro-phenol

  • Cas Number : 16292-90-3
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-amino-4-nitro-phenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79014
Availability details for
Quantity
  • Availability
  • Add to Cart