ADVH49B822E0 AChemBlock

3-amino-7,9-difluoro-1,3,4,5-tetrahydro-1-benzazepin-2-one

  • Cas Number : 1622853-51-3
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-amino-7-9-difluoro-1-3-4-5-tetrahydro-1-benzazepin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79046
Availability details for
Quantity
  • Availability
  • Add to Cart