ADVH49B822E1 AChemBlock

4-bromo-4'-tert-butylbiphenyl

  • Cas Number : 162258-89-1
  • Molecular Weight : 289.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-4-tert-butylbiphenyl.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71642
Availability details for
Quantity
  • Availability
  • Add to Cart