ADVH49B822E3 AChemBlock

exo-N,N-dimethyl-3-azabicyclo[3.1.0]hexan-6-amine dihydrochloride

  • Cas Number : 1622351-34-1
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/exo-n-n-dimethyl-3-azabicyclo-3-1-0-hexan-6-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79050
Availability details for
Quantity
  • Availability
  • Add to Cart