ADVH49B822E6 AChemBlock

4-Chloro-7-fluoro-6-nitroquinazoline

  • Cas Number : 162012-70-6
  • Molecular Weight : 227.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-fluoro-6-nitroquinazoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86673
Availability details for
Quantity
  • Availability
  • Add to Cart