ADVH49B822E8 AChemBlock

Benzamil hydrochloride

  • Cas Number : 161804-20-2
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/benzamil-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86594
Availability details for
Quantity
  • Availability
  • Add to Cart