ADVH49B822EE AChemBlock

methyl 3-bromopyrazolo[1,5-a]pyridine-5-carboxylate

  • Cas Number : 1610611-84-1
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-bromopyrazolo-1-5-a-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79108
Availability details for
Quantity
  • Availability
  • Add to Cart