ADVH49B822EF AChemBlock

6-bromo-8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine

  • Cas Number : 1610045-39-0
  • Molecular Weight : 247.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-8-chloro-3-methyl-1-2-4-triazolo-4-3-a-pyrazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79110
Availability details for
Quantity
  • Availability
  • Add to Cart