ADVH49B822F3 AChemBlock

(6-methoxy-3-pyridyl)hydrazine

  • Cas Number : 160664-95-9
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methoxy-3-pyridyl-hydrazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79142
Availability details for
Quantity
  • Availability
  • Add to Cart