ADVH49B822F9 AChemBlock

2,5-dichloropyrido[3,4-b]pyrazine

  • Cas Number : 1600511-80-5
  • Molecular Weight : 200.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-5-dichloropyrido-3-4-b-pyrazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79165
Availability details for
Quantity
  • Availability
  • Add to Cart