ADVH49B822FC AChemBlock

1-(5-bromopyridin-2-yl)ethan-1-ol

  • Cas Number : 159533-68-3
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-5-bromopyridin-2-yl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79184
Availability details for
Quantity
  • Availability
  • Add to Cart