ADVH49B822FE AChemBlock

6-fluoro-3-methyl-1H-indazole

  • Cas Number : 159305-16-5
  • Molecular Weight : 150.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-3-methyl-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79191
Availability details for
Quantity
  • Availability
  • Add to Cart