ADVH49B822FF AChemBlock

ethyl 4-(4-pyridyl)-1H-pyrazole-3-carboxylate

  • Cas Number : 1583555-40-1
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-4-pyridyl-1h-pyrazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79219
Availability details for
Quantity
  • Availability
  • Add to Cart