ADVH49B82302 AChemBlock

2-(2,2-difluoro-2H-1,3-benzodioxol-4-yl)acetonitrile

  • Cas Number : 157437-24-6
  • Molecular Weight : 197.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-difluoro-2h-1-3-benzodioxol-4-yl-acetonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79237
Availability details for
Quantity
  • Availability
  • Add to Cart