ADVH49B8230C AChemBlock

diethyl 1,4-dioxaspiro[4.4]nonane-8,8-dicarboxylate

  • Cas Number : 1557247-15-0
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/diethyl-1-4-dioxaspiro-4-4-nonane-8-8-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79315
Availability details for
Quantity
  • Availability
  • Add to Cart