ADVH49B8230F AChemBlock

7-bromo-2,3-dihydropyrazolo[5,1-b]oxazole

  • Cas Number : 1554086-90-6
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-3-dihydropyrazolo-5-1-b-oxazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79370
Availability details for
Quantity
  • Availability
  • Add to Cart