ADVH49B82310 AChemBlock

6-cyclopropyl-3-nitro-pyridin-2-amine

  • Cas Number : 1552286-03-9
  • Molecular Weight : 179.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-cyclopropyl-3-nitro-pyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79385
Availability details for
Quantity
  • Availability
  • Add to Cart