ADVH49B82313 AChemBlock

1-methyl-5-(propan-2-yl)-1H-pyrazole-3-carbaldehyde

  • Cas Number : 154927-04-5
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-5-propan-2-yl-1h-pyrazole-3-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79415
Availability details for
Quantity
  • Availability
  • Add to Cart