ADVH49B82316 AChemBlock

tert-butyl N-{[1-(bromomethyl)cyclopropyl]methyl}carbamate

  • Cas Number : 1545706-29-3
  • Molecular Weight : 264.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1-bromomethyl-cyclopropyl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79458
Availability details for
Quantity
  • Availability
  • Add to Cart