ADVH49B8231B AChemBlock

(3R,4R)-4-aminooxolan-3-ol

  • Cas Number : 153610-11-8
  • Molecular Weight : 103.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-4r-4-aminooxolan-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79519
Availability details for
Quantity
  • Availability
  • Add to Cart