ADVH49B8231E AChemBlock

(1-Benzyl-1,2,3,6-tetrahydropyridin-4-yl)methanamine

  • Cas Number : 153196-51-1
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-1-2-3-6-tetrahydropyridin-4-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86643
Availability details for
Quantity
  • Availability
  • Add to Cart