ADVH49B82323 AChemBlock

3-methyl-1H,4H,5H,6H-pyrrolo[3,4-c]pyrazole dihydrochloride

  • Cas Number : 1523617-96-0
  • Molecular Weight : 196.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1h-4h-5h-6h-pyrrolo-3-4-c-pyrazole-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79560
Availability details for
Quantity
  • Availability
  • Add to Cart